Learn More
ORS Nasco GLV DRY BOX BUTYL 15MIL 1PR
Catalog No. 502551659
Encompass_Preferred
Honeywell part# 8B1532/9Q. Honeywell Butyl Glove Box Gloves 8 in. Port Black Hand: Hand specific Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 9.75 Thickness: 15 mil 0.38 mm Circumference Hand: 9.75 in. 25 cm Diameter Cuff: 8 in. 20.3 cm Exterior Finish: Smooth Grip Design: Smooth Length: 32 in. 81.3 cm
Catalog No. 50-255-1659 Supplier ORS Nasco Supplier No. 0688B1532/9Q
Honeywell part# 8B1532/9Q. Honeywell Butyl Glove Box Gloves 8 in. Port Black Hand: Hand specific Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 9.75 Thickness: 15 mil 0.38 mm Circumference Hand: 9.75 in. 25 cm Diameter Cuff: 8 in. 20.3 cm Exterior Finish: Smooth Grip Design: Smooth Length: 32 in. 81.3 cm