Learn More
MSC Honeywell North Butyl Gloves, 15 mil, 32 in, 1 Pair
Catalog No. 502370353
Encompass
These butyl glovebox gloves are engineered for controlled environments, providing superior permeation resistance against vapors, gases, and a broad spectrum of chemicals. They are ideal for handling highly toxic substances in pharmaceutical, biotechnical, and nuclear applications.
  • Highest permeation resistance to vapors and gases
  • Resistant to ketones, esters, ozone, and oxidizing chemicals
  • Soft and flexible feel reduces hand fatigue at low temperatures
  • Complies with USDA and FDA regulations for food contact
  • Anti-static, latex-free, and silicone-free
Catalog No. 50-237-0353 Supplier MSC Supplier No. 86551637
These butyl glovebox gloves are engineered for controlled environments, providing superior permeation resistance against vapors, gases, and a broad spectrum of chemicals. They are ideal for handling highly toxic substances in pharmaceutical, biotechnical, and nuclear applications.
  • Highest permeation resistance to vapors and gases
  • Resistant to ketones, esters, ozone, and oxidizing chemicals
  • Soft and flexible feel reduces hand fatigue at low temperatures
  • Complies with USDA and FDA regulations for food contact
  • Anti-static, latex-free, and silicone-free