Learn More
Refrigiwear Inc. 0283R Regular Black XLarge Insulated High Dexterity
SDP
Catalog No. NC2742228
Encompass_Preferred
Warmth is more than just insulation; it's also staying dry and flexible to promote circulation. The Waterproof Abrasion Safety Glove features a waterproof barrier, along with fiberfill and foam insulation and a brushed lining, to help keep your hands dry and warm. It also boasts a neoprene and spandex back to give your hands plenty of flexibility while PVC abrasion pads on fingertips and palm protect you from the hazards of the job. Comfort Rating: -20�F Fiberfill plus foam insulation with brushed tricot lining Neoprene and spandex back with synthetic leather palm PVC abrasion pads on fingertips and palm Waterproof barrier Safety cuff with heavy-duty knit wrist  
Catalog No. NC2742228 Supplier Refrigiwear Inc. Supplier No. 0283RBLKXLG
Warmth is more than just insulation; it's also staying dry and flexible to promote circulation. The Waterproof Abrasion Safety Glove features a waterproof barrier, along with fiberfill and foam insulation and a brushed lining, to help keep your hands dry and warm. It also boasts a neoprene and spandex back to give your hands plenty of flexibility while PVC abrasion pads on fingertips and palm protect you from the hazards of the job. Comfort Rating: -20�F Fiberfill plus foam insulation with brushed tricot lining Neoprene and spandex back with synthetic leather palm PVC abrasion pads on fingertips and palm Waterproof barrier Safety cuff with heavy-duty knit wrist  