Learn More
MSC Honeywell North Chemical Resistant Gloves, Nitrile, X-Large, Supported, 1 Pair
Catalog No. 502370360
Encompass
The Honeywell North NK803 Nitri-Knit series of supported gloves are constructed from Nitrile over an Interlock knitted liner. This design enhances wearer comfort and provides improved resistance to tears and abrasions. The hand portion of the glove is treated with an anti-bacterial and anti-mildew agent, making them suitable for general handling tasks, especially those involving solvents, caustics, and animal fats.
  • Enhanced comfort due to interlock knitted liner
  • Improved resistance to tears and abrasions
  • Anti-bacterial and anti-mildew treated hand portion
  • Ideal for handling solvents, caustics, and animal fats
  • Chemical resistant
  • Resistant to cuts, snags, and punctures
  • Temperature range: -23°C to 149°C (-10°F to 300°F)
Catalog No. 50-237-0360 Supplier MSC Supplier No. 02016640
The Honeywell North NK803 Nitri-Knit series of supported gloves are constructed from Nitrile over an Interlock knitted liner. This design enhances wearer comfort and provides improved resistance to tears and abrasions. The hand portion of the glove is treated with an anti-bacterial and anti-mildew agent, making them suitable for general handling tasks, especially those involving solvents, caustics, and animal fats.
  • Enhanced comfort due to interlock knitted liner
  • Improved resistance to tears and abrasions
  • Anti-bacterial and anti-mildew treated hand portion
  • Ideal for handling solvents, caustics, and animal fats
  • Chemical resistant
  • Resistant to cuts, snags, and punctures
  • Temperature range: -23°C to 149°C (-10°F to 300°F)