Learn More
ORS Nasco G3 NXT* Nitrile Gloves, Large, 1000/case
Catalog No. NC1832088
Encompass_Preferred
Features textured fingertips, beaded cuff and 12" length. Contains no natural rubber latex. Static dissipative in use. Recommended for ISO Class 3 or higher cleanroom. Packaged in a Class 100 Cleanroom. AQL level of 1.5 for pinholes. White. 10 Bx/Cs.
This item is not returnable. View return policy
Catalog No. NC1832088 Supplier ORS Nasco Supplier No. 62993/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Features textured fingertips, beaded cuff and 12" length. Contains no natural rubber latex. Static dissipative in use. Recommended for ISO Class 3 or higher cleanroom. Packaged in a Class 100 Cleanroom. AQL level of 1.5 for pinholes. White. 10 Bx/Cs.