Learn More
ORS Nasco G3 NXT* Nitrile Gloves, Medium, 1000/case
Catalog No. NC1834707
Encompass_Preferred
Features textured fingertips, beaded cuff and 12" length. Contains no natural rubber latex. Static dissipative in use. Recommended for ISO Class 3 or higher cleanroom. Packaged in a Class 100 Cleanroom. AQL level of 1.5 for pinholes. White. 10 Bx/Cs
Catalog No. NC1834707 Supplier ORS Nasco Supplier No. 62992/DEL
Features textured fingertips, beaded cuff and 12" length. Contains no natural rubber latex. Static dissipative in use. Recommended for ISO Class 3 or higher cleanroom. Packaged in a Class 100 Cleanroom. AQL level of 1.5 for pinholes. White. 10 Bx/Cs