Learn More
RS Hughes Inc GLV VERSATOUCH 23-202 SZ9 1PR
SDP
Catalog No. NC1499653
Encompass_Preferred
Brand: AnsellTrade Name: VersaTouchSeries: 23-202Glove Material: PVCCoating Material: PVCSurface Finish: SmoothLining Material: AcrylicGender: UnisexSize: 9Protective Properties: Chemical-ResistantColor: BlueStandards Met: 21 CFR; EN 374; EN 511 021
This item is not returnable. View return policy
Catalog No. NC1499653 Supplier RS Hughes Inc Supplier No. 07649024619
Brand: AnsellTrade Name: VersaTouchSeries: 23-202Glove Material: PVCCoating Material: PVCSurface Finish: SmoothLining Material: AcrylicGender: UnisexSize: 9Protective Properties: Chemical-ResistantColor: BlueStandards Met: 21 CFR; EN 374; EN 511 021