Learn More
RS Hughes Inc GLV VERSATOUCH 23202 SZ10 1PR
SDP
Catalog No. NC1499654
Encompass_Preferred
Brand: AnsellTrade Name: VersaTouchSeries: 23-202Glove Material: PVCCoating Material: PVCSurface Finish: SmoothLining Material: AcrylicGender: UnisexSize: 10Protective Properties: Chemical-ResistantColor: BlueStandards Met: 21 CFR; EN 374; EN 511 021
This item is not returnable. View return policy
Catalog No. NC1499654 Supplier RS Hughes Inc Supplier No. 076409024620
Brand: AnsellTrade Name: VersaTouchSeries: 23-202Glove Material: PVCCoating Material: PVCSurface Finish: SmoothLining Material: AcrylicGender: UnisexSize: 10Protective Properties: Chemical-ResistantColor: BlueStandards Met: 21 CFR; EN 374; EN 511 021