Learn More
Abcam GL x -351322, 25MG
Catalog No. 502671222
Encompass_Preferred
MW 431.5 Da, Purity >=98%. GLX-351322 is a potent NADPH oxidase-4 (NOX-4) inhibitor. Displays selectivity for NOX-4 over NOX-2. GLX-351322 inhibits hydrogen peroxide production from tetracycline inducible NOX4-overexpressing cells (IC₅₀ = 5 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1222 Supplier Abcam Supplier No. AB28701025MG
MW 431.5 Da, Purity >=98%. GLX-351322 is a potent NADPH oxidase-4 (NOX-4) inhibitor. Displays selectivity for NOX-4 over NOX-2. GLX-351322 inhibits hydrogen peroxide production from tetracycline inducible NOX4-overexpressing cells (IC₅₀ = 5 μM).
The product is subject to the following: Abcam Restricted Use Statement