Learn More
American Radiolabeled Chemicals Inc GLY-3-PO4-2-3H 1LC
SDP
Catalog No. 5090104327
Encompass_Preferred
Glycerol, L-2-3H(N)-3-phosphate disodium salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 216.1, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-04327 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 02191MC
Add to Cart
Add to Cart
Request a Quote
Glycerol, L-2-3H(N)-3-phosphate disodium salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 20-30 Ci/mmol, M.W. 216.1, Shipped in Dry Ice, Exclusive