Learn More
Ubpbio Used for synthesis of Ub, SUMO or other Ub-like proteins with AMC conjugated at the C-terminus
Catalog No. 502339391
Encompass_Preferred
Gly-AMC can be used to synthesize Ub-AMC, SUMO-AMC, NEDD8-AMC and ISG15-AMC via the intein-mediated protein ligation technology.
Catalog No. 50-233-9391 Supplier Ubpbio Supplier No. G4110
Gly-AMC can be used to synthesize Ub-AMC, SUMO-AMC, NEDD8-AMC and ISG15-AMC via the intein-mediated protein ligation technology.