Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gly-Arg-AMC | 65147-19-5 | 99.37% | 388.42 | 10 MG
SDP

Catalog No. 5000399738
Encompass_Preferred

Gly-Arg-AMC is a peptide substrate of DPAP1, intended for research use only. This compound is characterized by its molecular properties and solubility for laboratory applications.

  • Peptide substrate of DPAP1.
  • Molecular weight of 388.42.
  • Chemical formula C18H24N6O4.
  • Appears as a solid.
  • Color ranges from white to off-white.
  • Sequence Gly-Arg-{AMC} (GR-{AMC}).
  • Soluble in DMSO and H2O.
  • Requires sealed storage, away from moisture and light.

Catalog No. 50-003-99738 Supplier Medchemexpress LLC Supplier No. HYP434210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gly-Arg-AMC is a peptide substrate of DPAP1, intended for research use only. This compound is characterized by its molecular properties and solubility for laboratory applications.

  • Peptide substrate of DPAP1.
  • Molecular weight of 388.42.
  • Chemical formula C18H24N6O4.
  • Appears as a solid.
  • Color ranges from white to off-white.
  • Sequence Gly-Arg-{AMC} (GR-{AMC}).
  • Soluble in DMSO and H2O.
  • Requires sealed storage, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.