Learn More
Medchemexpress LLC Gly-Arg-AMC | 65147-19-5 | 99.4% | 388.42 | 5 MG
SDP
Catalog No. 5000399598
Encompass_Preferred
Gly-Arg-AMC is a high-purity peptide substrate specifically designed for studying Dipeptidyl Aminopeptidase 1 (DPAP1) activity. It is provided in a solid, white to off-white appearance and is soluble in both DMSO and water. This substrate is essential for biochemical assays and research involving DPAP1, with specific storage recommendations to ensure long-term stability.
  • Functions as a specific peptide substrate for DPAP1
  • Supplied in a solid, white to off-white form
  • High purity
  • Soluble in DMSO and water
  • Requires sealed storage away from moisture and light for optimal stability
  • Stable as a powder at -80°C for 2 years or -20°C for 1 year
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-99598 Supplier Medchemexpress LLC Supplier No. HYP43425MG
Gly-Arg-AMC is a high-purity peptide substrate specifically designed for studying Dipeptidyl Aminopeptidase 1 (DPAP1) activity. It is provided in a solid, white to off-white appearance and is soluble in both DMSO and water. This substrate is essential for biochemical assays and research involving DPAP1, with specific storage recommendations to ensure long-term stability.
  • Functions as a specific peptide substrate for DPAP1
  • Supplied in a solid, white to off-white form
  • High purity
  • Soluble in DMSO and water
  • Requires sealed storage away from moisture and light for optimal stability
  • Stable as a powder at -80°C for 2 years or -20°C for 1 year
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month