Learn More
Medchemexpress LLC HY-P0295 2mg Medchemexpress, Gly-Arg-Gly-Asp-Ser CAS:96426-21-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02952MG
Encompass_Preferred
Medchemexpress, HY-P0295 2mg Gly-Arg-Gly-Asp-Ser CAS:96426-21-0 Gly-Arg-Gly-Asp-Ser is a pentapeptide that forms the cell-binding domain of a glycoprotein, osteopontin. Gly-Arg-Gly-Asp-Ser binds to integrin receptors αvβ3 and αvβ5 with estimated IC 50 of ∼5 and ∼6.5 μM [1] Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4103