Learn More
Medchemexpress LLC Gly-Arg-Gly-Asp-Ser (GRGDS) | 96426-21-0 | 99.4% | 490.47 | 25 MG
SDP
Catalog No. 5000426167
Encompass_Preferred
Gly-Arg-Gly-Asp-Ser (GRGDS) is a pentapeptide that forms the cell-binding domain of osteopontin, a glycoprotein. It binds to integrin receptors αvβ3 and αvβ5 with estimated IC50 of ~5 and ~6.5 μM.
  • Binds to integrin receptors αvβ3 and αvβ5.
  • Exhibits antagonist activity at the integrin alpha4beta1 receptor in human CCRF-CEM cells.
  • Inhibits integrin alphaVbeta1-mediated CHO cell adhesion to fibronectin.
  • Enhances osteoblast-like cell adhesion, spreading, and proliferation.
  • Primarily intended for research purposes.
Catalog No. 50-004-26167 Supplier Medchemexpress LLC Supplier No. 25MGHYP0295
Gly-Arg-Gly-Asp-Ser (GRGDS) is a pentapeptide that forms the cell-binding domain of osteopontin, a glycoprotein. It binds to integrin receptors αvβ3 and αvβ5 with estimated IC50 of ~5 and ~6.5 μM.
  • Binds to integrin receptors αvβ3 and αvβ5.
  • Exhibits antagonist activity at the integrin alpha4beta1 receptor in human CCRF-CEM cells.
  • Inhibits integrin alphaVbeta1-mediated CHO cell adhesion to fibronectin.
  • Enhances osteoblast-like cell adhesion, spreading, and proliferation.
  • Primarily intended for research purposes.