Learn More
Meridian Life Science Gly-Free DNA Helicase -HC, 200UL
SDP
Catalog No. 5000856303
Encompass_Preferred
Glycerol-Free DNA Helicase (HC) Kit is a multi-domain enzyme with both ATPase and helicase activities. It unwinds double-stranded DNA with 3’ to 5’ with respect to the bound strand and initiates unwinding most effectively when a single-stranded region is present. Helicase works in coordination with an accessory protein, methyl-directed mismatch repair (MutL) protein that stimulates the helicase activity. Glycerol-Free DNA Helicase (HC) has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization. This gives you the flexibility to confidently store mixtures at ambient temperature or produce diagnostic assays that rely on miniaturized reaction components, such as point-of-care assays.
Catalog No. 50-008-56303 Supplier Meridian Life Science Supplier No. MDX24102
Glycerol-Free DNA Helicase (HC) Kit is a multi-domain enzyme with both ATPase and helicase activities. It unwinds double-stranded DNA with 3’ to 5’ with respect to the bound strand and initiates unwinding most effectively when a single-stranded region is present. Helicase works in coordination with an accessory protein, methyl-directed mismatch repair (MutL) protein that stimulates the helicase activity. Glycerol-Free DNA Helicase (HC) has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization. This gives you the flexibility to confidently store mixtures at ambient temperature or produce diagnostic assays that rely on miniaturized reaction components, such as point-of-care assays.