Learn More
Meridian Life Science Gly-Free DNA Pol I Klenow, 1ML
SDP
Catalog No. 5000856380
Encompass_Preferred
Glycerol-Free DNA Pol I Klenow Fragment (HC) is a high concentration enzyme (50 U/uL) and is mainly used in next-generation sequencing (NGS) library preparation, where it can be used separately or as part of an End-Repair Mix, for double stranded DNA blunting by 3’ overhang removal and fill-in of 5’ overhangs for subsequent ligation of adaptors by T4 DNA Ligase. The enzyme displays moderate strand displacement activity and can also be used to generate probes using random primers, dideoxy sequencing and random priming labelling. Glycerol-Free DNA Pol I Klenow Fragment (HC) is in a glycerol-free storage buffer and is supplied with a Lyo-Ready Klenow Reaction Buffer and has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization.
Catalog No. 50-008-56380 Supplier Meridian Life Science Supplier No. MDX2081
Glycerol-Free DNA Pol I Klenow Fragment (HC) is a high concentration enzyme (50 U/uL) and is mainly used in next-generation sequencing (NGS) library preparation, where it can be used separately or as part of an End-Repair Mix, for double stranded DNA blunting by 3’ overhang removal and fill-in of 5’ overhangs for subsequent ligation of adaptors by T4 DNA Ligase. The enzyme displays moderate strand displacement activity and can also be used to generate probes using random primers, dideoxy sequencing and random priming labelling. Glycerol-Free DNA Pol I Klenow Fragment (HC) is in a glycerol-free storage buffer and is supplied with a Lyo-Ready Klenow Reaction Buffer and has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization.