Learn More
Meridian Life Science Gly-Free DNA Pol I Klenow exo-, 50UL
SDP
Catalog No. 5000856352
Encompass_Preferred
Glycerol-Free DNA Pol I Klenow Fragment exo- (HC) is a high concentration enzyme (50 U/uL) has mutations (D355A, E357A) which eliminates the 3’-5’ exonuclease activity. It is designed for dA and dT tailing for adapter ligation in next generation sequencing library preparation. The enzyme displays moderate strand displacement activity and can also be used to generate probes using random primers, dideoxy sequencing and random priming labelling. Glycerol-Free DNA Pol I Klenow Fragment exo- (HC) is in a glycerol-free storage buffer and is supplied with a Lyo-Ready Klenow Reaction Buffer and has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization.
Catalog No. 50-008-56352 Supplier Meridian Life Science Supplier No. MDX218005
Glycerol-Free DNA Pol I Klenow Fragment exo- (HC) is a high concentration enzyme (50 U/uL) has mutations (D355A, E357A) which eliminates the 3’-5’ exonuclease activity. It is designed for dA and dT tailing for adapter ligation in next generation sequencing library preparation. The enzyme displays moderate strand displacement activity and can also be used to generate probes using random primers, dideoxy sequencing and random priming labelling. Glycerol-Free DNA Pol I Klenow Fragment exo- (HC) is in a glycerol-free storage buffer and is supplied with a Lyo-Ready Klenow Reaction Buffer and has been optimized to deliver excellent, stable performance without glycerol, allowing you to deliver reliable results with the added capability for lyophilization.