Learn More
Meridian Life Science Gly-Free High-Fidelity Pfu -HC, 25UL
SDP
Catalog No. 5000856301
Encompass_Preferred
Glycerol-Free High-Fidelity Pfu (HC) is a high-fidelity, thermostable DNA-dependent DNA polymerase from Pyrococcus furiosus. The 3’ - 5’ proofreading exonuclease activity of Pfu polymerase makes it suitable for applications that require high-fidelity DNA synthesis or blunt-ended PCR fragments. GlycerolFree High-Fidelity Pfu (HC) is supplied in a glycerol-free storage buffer and is accompanied by a 5x Reaction Buffer that contains magnesium, dNTPs and excipients required for lyophilization. Glycerol-Free High-Fidelity Pfu (HC) enables flexible and scalable reaction volumes, and is suitable for applications such NGS library amplification, gene expression, cloning and site-directed mutagenesis.
Catalog No. 50-008-56301 Supplier Meridian Life Science Supplier No. MDX2030025
Glycerol-Free High-Fidelity Pfu (HC) is a high-fidelity, thermostable DNA-dependent DNA polymerase from Pyrococcus furiosus. The 3’ - 5’ proofreading exonuclease activity of Pfu polymerase makes it suitable for applications that require high-fidelity DNA synthesis or blunt-ended PCR fragments. GlycerolFree High-Fidelity Pfu (HC) is supplied in a glycerol-free storage buffer and is accompanied by a 5x Reaction Buffer that contains magnesium, dNTPs and excipients required for lyophilization. Glycerol-Free High-Fidelity Pfu (HC) enables flexible and scalable reaction volumes, and is suitable for applications such NGS library amplification, gene expression, cloning and site-directed mutagenesis.