Learn More
Meridian Life Science Gly-Free HS Tth DNA Polymerase, 20UL
SDP
Catalog No. 5000856449
Encompass_Preferred
Glycerol-Free Tth DNA Polymerase (HC) is a high concentration thermostable DNA polymerase with intrinsic reverse transcriptase (RT) activity. The elevated temperatures of Tth DNA Polymerase activity overcomes the problems posed by RNA secondary structure. Resulting cDNA can be amplified by PCR using the same enzyme in the presence of Mg2+ ions. The ability of Tth DNA Polymerase to perform both reverse transcription and DNA amplification at elevated temperatures allows this enzyme to be used for quantitative RT-PCR, cloning and gene expression analysis of cellular and viral RNA. It is supplied with a 5x Reaction Buffer that contains dNTPs and excipients required for lyophilization.
Catalog No. 50-008-56449 Supplier Meridian Life Science Supplier No. MDX205002
Glycerol-Free Tth DNA Polymerase (HC) is a high concentration thermostable DNA polymerase with intrinsic reverse transcriptase (RT) activity. The elevated temperatures of Tth DNA Polymerase activity overcomes the problems posed by RNA secondary structure. Resulting cDNA can be amplified by PCR using the same enzyme in the presence of Mg2+ ions. The ability of Tth DNA Polymerase to perform both reverse transcription and DNA amplification at elevated temperatures allows this enzyme to be used for quantitative RT-PCR, cloning and gene expression analysis of cellular and viral RNA. It is supplied with a 5x Reaction Buffer that contains dNTPs and excipients required for lyophilization.