Learn More
Meridian Life Science Gly-Free HS Tth DNA Polymerase, 500UL
SDP
Catalog No. 5000856350
Encompass_Preferred
Glycerol-Free Tth DNA Polymerase (HC) is a high concentration thermostable DNA polymerase with intrinsic reverse transcriptase (RT) activity. The elevated temperatures of Tth DNA Polymerase activity overcomes the problems posed by RNA secondary structure. Resulting cDNA can be amplified by PCR using the same enzyme in the presence of Mg2+ ions. The ability of Tth DNA Polymerase to perform both reverse transcription and DNA amplification at elevated temperatures allows this enzyme to be used for quantitative RT-PCR, cloning and gene expression analysis of cellular and viral RNA. It is supplied with a 5x Reaction Buffer that contains dNTPs and excipients required for lyophilization.
Catalog No. 50-008-56350 Supplier Meridian Life Science Supplier No. MDX20505
Glycerol-Free Tth DNA Polymerase (HC) is a high concentration thermostable DNA polymerase with intrinsic reverse transcriptase (RT) activity. The elevated temperatures of Tth DNA Polymerase activity overcomes the problems posed by RNA secondary structure. Resulting cDNA can be amplified by PCR using the same enzyme in the presence of Mg2+ ions. The ability of Tth DNA Polymerase to perform both reverse transcription and DNA amplification at elevated temperatures allows this enzyme to be used for quantitative RT-PCR, cloning and gene expression analysis of cellular and viral RNA. It is supplied with a 5x Reaction Buffer that contains dNTPs and excipients required for lyophilization.