Learn More
Meridian Life Science Gly-Free Non-Bac Taq HS Poly, 20UL
SDP
Supplier:  Meridian Life Science MDX252002
Encompass_Preferred
Glycerol-Free Non-Bac HS Taq Polymerase (HC) is a highly thermostable DNA polymerase derived from the thermophilic bacterium Thermus aquaticus, pre-mixed with a Taq Hot-start antibody and produced using a recombinant eukaryotic expression system. This unique production method ensures the enzyme is completely free of E. coli DNA contamination, making it the ideal choice for sensitive PCR applications where even trace levels of bacterial DNA could compromise assay accuracy. In addition, Glycerol-Free Non-Bac HS Taq Polymerase (HC) is fully compatible with the production of lyophilized or air-dried qPCR and RT-qPCR reagents that are stable at ambient temperatures.
Catalog No. 50-008-56477