Learn More
Meridian Life Science Gly-Free Phi29 Polymerase -HC, 20UL
SDP
Catalog No. 5000856537
Encompass_Preferred
Glycerol-Free Phi29 DNA Polymerase (HC) is a highly processive DNA-dependent DNA polymerase with strong strand displacement activity, which enables efficient isothermal DNA amplification from low DNA inputs. Glycerol-Free Phi29 DNA Polymerase (HC) 3′ - 5′ exonuclease activity delivers high fidelity amplification, making it an appropriate solution for sequencing DNA template preparation. It is supplied with a 5x Reaction Buffer that contains excipients required for lyophilization. Glycerol-Free Phi29 DNA Polymerase (HC) enables flexible and scalable reaction volumes, and is suitable for applications such as rolling circle amplification (RCA) and multiple displacement amplification (MDA).
Catalog No. 50-008-56537 Supplier Meridian Life Science Supplier No. MDX240002
Glycerol-Free Phi29 DNA Polymerase (HC) is a highly processive DNA-dependent DNA polymerase with strong strand displacement activity, which enables efficient isothermal DNA amplification from low DNA inputs. Glycerol-Free Phi29 DNA Polymerase (HC) 3′ - 5′ exonuclease activity delivers high fidelity amplification, making it an appropriate solution for sequencing DNA template preparation. It is supplied with a 5x Reaction Buffer that contains excipients required for lyophilization. Glycerol-Free Phi29 DNA Polymerase (HC) enables flexible and scalable reaction volumes, and is suitable for applications such as rolling circle amplification (RCA) and multiple displacement amplification (MDA).