Learn More
Meridian Life Science Gly-Free T4 DNA Polymerase, 250UL
SDP
Catalog No. 5000856188
Encompass_Preferred
A repair enzyme that converts DNA or RNA 3′ PO4/5′ OH ends into 3′ OH/5′ PO4 ends, which are then suitable for ligation.
Catalog No. 50-008-56188 Supplier Meridian Life Science Supplier No. MDX206025
A repair enzyme that converts DNA or RNA 3′ PO4/5′ OH ends into 3′ OH/5′ PO4 ends, which are then suitable for ligation.