Learn More
Meridian Life Science Gly-Free T4 DNA Polymerase, 50UL
SDP
Catalog No. 5000856213
Encompass_Preferred
Glycerol-Free T4 DNA Polymerase (HC) is a DNA-dependent DNA polymerase that catalyzes 5- 3´ synthesis of DNA from primed single-stranded DNA template. Glycerol-Free T4 DNA Polymerase (HC) possesses a strong 3- 5 exonuclease (proofreading) activity, but lacks 5-3 exonuclease activity. It is supplied with a 5x Reaction Buffer that contains excipients required for lyophilization. Glycerol-Free T4 DNA Polymerase (HC) enables flexible and scalable reaction volumes and is suitable for applications such as next-generation sequencing (NGS) library preparation, where it can be used separately or as part of an End-Repair Mix to blunt DNA ends (fill-in 5’-overhangs or/and removal of 3’-overhangs) or for probe labelling.
Catalog No. 50-008-56213 Supplier Meridian Life Science Supplier No. MDX207005
Glycerol-Free T4 DNA Polymerase (HC) is a DNA-dependent DNA polymerase that catalyzes 5- 3´ synthesis of DNA from primed single-stranded DNA template. Glycerol-Free T4 DNA Polymerase (HC) possesses a strong 3- 5 exonuclease (proofreading) activity, but lacks 5-3 exonuclease activity. It is supplied with a 5x Reaction Buffer that contains excipients required for lyophilization. Glycerol-Free T4 DNA Polymerase (HC) enables flexible and scalable reaction volumes and is suitable for applications such as next-generation sequencing (NGS) library preparation, where it can be used separately or as part of an End-Repair Mix to blunt DNA ends (fill-in 5’-overhangs or/and removal of 3’-overhangs) or for probe labelling.