Learn More
Meridian Life Science Gly-Free Taq HS Poly 50 U/uL, 20UL
SDP
Catalog No. 5000856220
Encompass_Preferred
Glycerol-Free Taq HS 50U/uL is supplied as a separate 10x, high‑performance glycerol‑free Taq DNA Polymerase, glycerol‑free Taq antibody and glycerol‑free enzyme dilution buffer. Separating these components allows greater flexibility in assay development, making Glycerol-Free Taq HS 50 U/uL suitable for difficult, multiplex PCR that require fast amplification, whilst maintaining both high specificity and sensitivity. Glycerol-Free Taq HS 50U/uL is ideal for PCR kit manufactures and PCR molecular test facilities, requiring large amounts of DNA polymerases. The highly concentrated Glycerol-Free Taq HS 50U/uL allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.
Catalog No. 50-008-56220 Supplier Meridian Life Science Supplier No. MDX011002
Glycerol-Free Taq HS 50U/uL is supplied as a separate 10x, high‑performance glycerol‑free Taq DNA Polymerase, glycerol‑free Taq antibody and glycerol‑free enzyme dilution buffer. Separating these components allows greater flexibility in assay development, making Glycerol-Free Taq HS 50 U/uL suitable for difficult, multiplex PCR that require fast amplification, whilst maintaining both high specificity and sensitivity. Glycerol-Free Taq HS 50U/uL is ideal for PCR kit manufactures and PCR molecular test facilities, requiring large amounts of DNA polymerases. The highly concentrated Glycerol-Free Taq HS 50U/uL allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.