Learn More
Meridian Life Science Gly-Free Taq Poly 50 U/uL, 500UL
SDP
Catalog No. 5000856417
Encompass_Preferred
The highly concentrated Glycerol-Free Taq HS 50U/uL allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.
Catalog No. 50-008-56417 Supplier Meridian Life Science Supplier No. MDX01105A
The highly concentrated Glycerol-Free Taq HS 50U/uL allows for concentrated bulk production of PCR master mixes and the glycerol free format is useful when using automated pipetting robots and other applications where accurate pipetting of small volumes is crucial.