Learn More
Meridian Life Science Gly-Free Thermo Helicase -HC, 200UL
SDP
Catalog No. 5000856187
Encompass_Preferred
Glycerol-Free DNA Thermostable Helicase (HC) Kit is a multi-domain enzyme with both ATPase and helicase activities, without a requirement for an overhang structure. It functions with a wide range of DNA substrates and exhibits high thermal stability. It is supplied in a glycerol-free storage buffer and is accompanied by a 5x lyophilization compatible Reaction Buffer. Glycerol-Free Thermostable DNA Helicase (HC) facilitates flexible and scalable reaction volumes and is suitable for use in diagnostic assays that rely on miniaturized reaction components, such as point-of-care assays.
Catalog No. 50-008-56187 Supplier Meridian Life Science Supplier No. MDX24202
Glycerol-Free DNA Thermostable Helicase (HC) Kit is a multi-domain enzyme with both ATPase and helicase activities, without a requirement for an overhang structure. It functions with a wide range of DNA substrates and exhibits high thermal stability. It is supplied in a glycerol-free storage buffer and is accompanied by a 5x lyophilization compatible Reaction Buffer. Glycerol-Free Thermostable DNA Helicase (HC) facilitates flexible and scalable reaction volumes and is suitable for use in diagnostic assays that rely on miniaturized reaction components, such as point-of-care assays.