Learn More
Cayman Chemical Gly-Gly-AMC (trifluoroacetate salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502896308
Encompass_Preferred
A fluorogenic peptide substrate; used to assess the activity of bacterial proteases from P. aeruginosa and S. aureus; enzyme activity can be quantified by fluorescent detection of free AMC (also known as 7-amino-4-methylcoumarin), which is excited at 340-360 nm and emits at 440-460 nm
Catalog No. 50-289-6308 Supplier Cayman Chemical Supplier No. 851401 MG
A fluorogenic peptide substrate; used to assess the activity of bacterial proteases from P. aeruginosa and S. aureus; enzyme activity can be quantified by fluorescent detection of free AMC (also known as 7-amino-4-methylcoumarin), which is excited at 340-360 nm and emits at 440-460 nm