Learn More
Medchemexpress LLC HY-145066 25mg , Gly-Gly-Gly-PEG4-azide CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14506625MG
Encompass_Preferred
HY-145066 25mg Gly-Gly-Gly-PEG4-azide CAS: Gly-Gly-Gly-PEG4-azide is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1471