Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gly-gly-gly-PEG4-azide | 3070260-22-6 | 99.8% | 433.46 g/mol | C16H31N7O7 | 50 MG
SDP

Catalog No. 5000202558
Encompass_Preferred

Gly-gly-gly-PEG4-azide is a PEGylated tri-glycine linker bearing a terminal azide functional group used for bioconjugation and click chemistry. It facilitates attachment of payloads to peptides, proteins, or small molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC).

  • Enables CuAAC and SPAAC bioconjugation reactions
  • High purity: 99.8% as supplied
  • Molecular weight: 433.46 g/mol; formula C16H31N7O7
  • Typical applications: ADC linker chemistry and payload attachment
  • Recommended storage: -20 °C under inert gas; in solution: -80 °C (6 months)
  • Available as a 50 MG vial

Catalog No. 50-002-02558 Supplier Medchemexpress LLC Supplier No. HY14506650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gly-gly-gly-PEG4-azide is a PEGylated tri-glycine linker bearing a terminal azide functional group used for bioconjugation and click chemistry. It facilitates attachment of payloads to peptides, proteins, or small molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC).

  • Enables CuAAC and SPAAC bioconjugation reactions
  • High purity: 99.8% as supplied
  • Molecular weight: 433.46 g/mol; formula C16H31N7O7
  • Typical applications: ADC linker chemistry and payload attachment
  • Recommended storage: -20 °C under inert gas; in solution: -80 °C (6 months)
  • Available as a 50 MG vial

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.