Learn More
Medchemexpress LLC Gly-gly-gly-PEG4-DBCO | 2353409-80-8 | 95.1% | 694.77 g/mol | C35H46N6O9 | 10 MG
SDP
Catalog No. 5000203814
Encompass_Preferred
Gly-Gly-Gly-PEG4-DBCO is a four-unit polyethylene glycol (PEG4) linker terminated with a dibenzocyclooctyne (DBCO) group, intended for click-chemistry bioconjugation such as antibody-drug conjugate synthesis. It reacts with azide-functionalized molecules via strain-promoted alkyne-azide cycloaddition (SPAAC), and is supplied as a powder suitable for laboratory conjugation workflows.
  • Four-unit PEG linker with a DBCO functional group.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide partners.
  • Suitable for site-specific bioconjugation and antibody-drug conjugate synthesis.
  • Powder form with defined storage stability at low temperatures.
  • Analytically characterized with reported purity for experimental reproducibility.
Catalog No. 50-002-03814 Supplier Medchemexpress LLC Supplier No. HY14030910MG
Gly-Gly-Gly-PEG4-DBCO is a four-unit polyethylene glycol (PEG4) linker terminated with a dibenzocyclooctyne (DBCO) group, intended for click-chemistry bioconjugation such as antibody-drug conjugate synthesis. It reacts with azide-functionalized molecules via strain-promoted alkyne-azide cycloaddition (SPAAC), and is supplied as a powder suitable for laboratory conjugation workflows.
  • Four-unit PEG linker with a DBCO functional group.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide partners.
  • Suitable for site-specific bioconjugation and antibody-drug conjugate synthesis.
  • Powder form with defined storage stability at low temperatures.
  • Analytically characterized with reported purity for experimental reproducibility.