Learn More
Medchemexpress LLC Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH | 1599440-20-6 | 99.98% | C18H25N5O7 | 250 MG
SDP
Catalog No. 5000358349
Encompass_Preferred
Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is an ADC Linker that can be used to synthesize Drug-Linker Conjugates for Antibody-Drug Conjugates. It is particularly useful for the synthesis of Deruxtecan, a toxic drug-linker conjugate. This compound appears as a white to off-white solid and is intended for research use only.
  • Can be used to synthesize drug-linker conjugates for antibody-drug conjugates
  • Particularly useful for the synthesis of Deruxtecan
  • Appears as a white to off-white solid
  • Intended for research use only
Catalog No. 50-003-58349 Supplier Medchemexpress LLC Supplier No. 250MGHY45672
Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is an ADC Linker that can be used to synthesize Drug-Linker Conjugates for Antibody-Drug Conjugates. It is particularly useful for the synthesis of Deruxtecan, a toxic drug-linker conjugate. This compound appears as a white to off-white solid and is intended for research use only.
  • Can be used to synthesize drug-linker conjugates for antibody-drug conjugates
  • Particularly useful for the synthesis of Deruxtecan
  • Appears as a white to off-white solid
  • Intended for research use only