Learn More
Medchemexpress LLC Gly-Leu-Met-NH2 | 4652-64-6 | 99.8% | 318.44 | 10 MG
SDP
Catalog No. 5000398463
Encompass_Preferred
Gly-Leu-Met-NH2 is a C-terminal tripeptide of Substance P, a neuropeptide. This product appears as a white to off-white solid and is intended for research use only, not for patient use.
  • C-terminal tripeptide of Substance P
  • Appears as a white to off-white solid
  • For research use only
  • Soluble in DMSO at 100 mg/mL
  • Targeted to Neurokinin Receptor
  • Involved in GPCR/G Protein and neuronal signaling pathways
Catalog No. 50-003-98463 Supplier Medchemexpress LLC Supplier No. HYP388810MG
Gly-Leu-Met-NH2 is a C-terminal tripeptide of Substance P, a neuropeptide. This product appears as a white to off-white solid and is intended for research use only, not for patient use.
  • C-terminal tripeptide of Substance P
  • Appears as a white to off-white solid
  • For research use only
  • Soluble in DMSO at 100 mg/mL
  • Targeted to Neurokinin Receptor
  • Involved in GPCR/G Protein and neuronal signaling pathways