Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLY-LEU-TYR, MP Biomedicals™

Catalog No. ICN15726480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15726480 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15726480 Supplier MP Biomedicals, Inc Supplier No. 0215726480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4306-24-5
Molecular Formula C17H25N3O5
Molecular Weight (g/mol) 351.403
InChI Key AFWYPMDMDYCKMD-KBPBESRZSA-N
Synonym h-gly-leu-tyr-oh, gly-leu-tyr, glycyl-l-leucyl-l-tyrosine, chembl55636, gly-leu-tyr tlc, s-2-s-2-2-aminoacetamido-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-2-aminoacetamido-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-2-aminoacetyl amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid
PubChem CID 96817
IUPAC Name (2S)-2-[[(2S)-2-[(2-aminoacetyl)amino]-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)NC(=O)CN

Specifications

Quantity 0.1 g
Formula Weight 351.4
Physical Form Crystalline
Chemical Name or Material GLY-LEU-TYR
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.