Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin

Catalog No. MP3AFC03325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3AFC03325 25mg
MP3AFC03310 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP3AFC03325 Supplier MP Biomedicals™ Supplier No. 03AFC03325
Add to Cart
Add to Cart

Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder

  • Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
  • In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
  • It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Powder
For Use With (Application) Cell Migration Analysis; Cell Signaling Analysis
Quantity 25mg
Molecular Weight (g/mol) 433.38 ; 574 (Including TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Keep cool and dry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.