Learn More
Medchemexpress LLC Gly-Phe-AMC | 201852-70-2 | 98.8% | 379.41 | 1 MG
SDP
Catalog No. 5000399602
Encompass_Preferred
Gly-Phe-AMC is a fluorogenic peptide substrate composed of a peptide sequence of glycine and phenylalanine, linked to the fluorophore AMC. It also functions as a cathepsin C substrate and is widely used to detect the activity of various proteases.
  • Fluorogenic peptide substrate
  • Functions as a cathepsin C substrate
  • Widely used to detect the activity of various proteases
Catalog No. 50-003-99602 Supplier Medchemexpress LLC Supplier No. HYP44251MG
Gly-Phe-AMC is a fluorogenic peptide substrate composed of a peptide sequence of glycine and phenylalanine, linked to the fluorophore AMC. It also functions as a cathepsin C substrate and is widely used to detect the activity of various proteases.
  • Fluorogenic peptide substrate
  • Functions as a cathepsin C substrate
  • Widely used to detect the activity of various proteases