Learn More
Selleck Chemical LLC Gly-Phe b-naphthylamide
SDP
Catalog No. 5000002743
Encompass_Preferred
Gly-Phe -naphthylamide (Gly-Phe beta-naphthylamide) is a substrate of cathepsin C that accumulates within the lysosome Gly-Phe- -naphthylamide can inhibit the cathepsin-dependent activation of caspase-8
Catalog No. 50-000-02743 Supplier Selleck Chemical LLC Supplier No. S684625MG
Gly-Phe -naphthylamide (Gly-Phe beta-naphthylamide) is a substrate of cathepsin C that accumulates within the lysosome Gly-Phe- -naphthylamide can inhibit the cathepsin-dependent activation of caspase-8