Learn More
Enzo Life Sciences GP-AMC (5 mg) CAS: 115035-46-6
SDP
Catalog No. NC1692256
Encompass_Preferred
DPP substrate. Fluorogenic substrate for dipeptidyl peptidase IV (DPIV, DPPIV, CD26) (.in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em=460 nm. Alternative Name: Gly-Pro-AMC, Dipeptidyl peptidase substrate (fluorogenic). MW: 410.3. CAS: 115035-46-6. Purity: ≥97%. Sequence H-Gly-Pro-7-amino-4-methylcoumarin. Long Term Storage: -20°C
Catalog No. NC1692256 Supplier Enzo Life Sciences Supplier No. BMLP1890005
DPP substrate. Fluorogenic substrate for dipeptidyl peptidase IV (DPIV, DPPIV, CD26) (.in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em=460 nm. Alternative Name: Gly-Pro-AMC, Dipeptidyl peptidase substrate (fluorogenic). MW: 410.3. CAS: 115035-46-6. Purity: ≥97%. Sequence H-Gly-Pro-7-amino-4-methylcoumarin. Long Term Storage: -20°C