Learn More
Medchemexpress LLC Gly-Pro-AMC hydrobromide | 115035-46-6 | 98.68% | 410.26 | 50 MG
SDP
Catalog No. 5000350687
Encompass_Preferred
Gly-Pro-AMC hydrobromide is a fluorescent dye that can be used as a specific fluorescent substrate for detecting Dipeptidyl peptidase IV (DPP-IV) activity. It exhibits fluorescence properties with an excitation wavelength of 350 nm and fluorescence emission spectrum detection at 450 nm.
  • Specific fluorescent substrate for detecting DPP-IV activity
  • Fluorescence properties with excitation wavelength of 350 nm
  • Fluorescence emission spectrum detection at 450 nm
  • Can be used at 50 μM for 20 min at 37 °C
Catalog No. 50-003-50687 Supplier Medchemexpress LLC Supplier No. HY13783450MG
Gly-Pro-AMC hydrobromide is a fluorescent dye that can be used as a specific fluorescent substrate for detecting Dipeptidyl peptidase IV (DPP-IV) activity. It exhibits fluorescence properties with an excitation wavelength of 350 nm and fluorescence emission spectrum detection at 450 nm.
  • Specific fluorescent substrate for detecting DPP-IV activity
  • Fluorescence properties with excitation wavelength of 350 nm
  • Fluorescence emission spectrum detection at 450 nm
  • Can be used at 50 μM for 20 min at 37 °C