Learn More
Medchemexpress LLC Gly-Pro-Glu | 32302-76-4 | 99.6% | 301.30 g/mol | C12H19N3O6 | 50 MG
SDP
Catalog No. 5000228723
Encompass_Preferred
Gly-Pro-Glu is an N-terminal tripeptide derived from insulin-like growth factor-I with reported neuroactive effects, provided as a high-purity research-grade powder suitable for in vitro biochemical and pharmacological studies.
  • Neuroactive tripeptide that modulates acetylcholine release.
  • High purity (99.6%) suitable for analytical and bioassay applications.
  • Supplied as a powder in milligram-scale quantities for laboratory use.
  • Store sealed and protected from moisture and light; use frozen storage for long-term stability.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-28723 Supplier Medchemexpress LLC Supplier No. HY11748350MG
Gly-Pro-Glu is an N-terminal tripeptide derived from insulin-like growth factor-I with reported neuroactive effects, provided as a high-purity research-grade powder suitable for in vitro biochemical and pharmacological studies.
  • Neuroactive tripeptide that modulates acetylcholine release.
  • High purity (99.6%) suitable for analytical and bioassay applications.
  • Supplied as a powder in milligram-scale quantities for laboratory use.
  • Store sealed and protected from moisture and light; use frozen storage for long-term stability.
  • For research use only; not for human or clinical applications.