Learn More
Carbosynth LLC. Gly-Pro-MCA (5 mg vial), 1ea, MW: 329.35, Molecular formula: C17H19N3O4, CAS# 67341-42-8
SDP
Supplier:  Carbosynth LLC. MGP3090V5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Glycyl-L-Proline 4-Methyl-Coumaryl-7-Amide, Gly-Pro-AMC, GP-AMC, GP-MCA, Gly-Pro-NH-trifluoromethylcoumarin, Purity: , Grade AA, Application: Substrate for X-prolyl Dipeptidyl-Aminopeptidase IV (DPP-IV), Salt Form:, Tosylate Form, Storage Temperature: -20C, References: T. Kato, T. Nagatsu, T. Kimura, and S. Sakakibara, Biochem. Med., 19, 351 (1978).
Catalog No. 50-168-5977