Learn More
Medchemexpress LLC Gly-Pro-pNA hydrochloride | 103213-34-9 | 99.9% | 25 MG
SDP
Catalog No. 5000232332
Encompass_Preferred
Gly-Pro-pNA hydrochloride is a chromogenic substrate for dipeptidyl peptidase IV (DPP-IV). Upon enzymatic cleavage it releases p-nitroaniline, enabling colorimetric measurement of DPP-IV activity and inhibitor screening. Supplied as a solid, it is suitable for enzymatic assays and biochemical studies.
  • Chromogenic DPP-IV substrate releasing p-nitroaniline for colorimetric assays.
  • High purity (99.92%) suitable for biochemical assays.
  • Molecular weight 328.76 and formula C13H17ClN4O4.
  • Supplied as a solid in small-scale packaging for laboratory use.
  • Useful for screening DPP-IV inhibitors and enzymatic activity studies.
Catalog No. 50-002-32332 Supplier Medchemexpress LLC Supplier No. HY11924425MG
Gly-Pro-pNA hydrochloride is a chromogenic substrate for dipeptidyl peptidase IV (DPP-IV). Upon enzymatic cleavage it releases p-nitroaniline, enabling colorimetric measurement of DPP-IV activity and inhibitor screening. Supplied as a solid, it is suitable for enzymatic assays and biochemical studies.
  • Chromogenic DPP-IV substrate releasing p-nitroaniline for colorimetric assays.
  • High purity (99.92%) suitable for biochemical assays.
  • Molecular weight 328.76 and formula C13H17ClN4O4.
  • Supplied as a solid in small-scale packaging for laboratory use.
  • Useful for screening DPP-IV inhibitors and enzymatic activity studies.