Learn More
Cayman Chemical Gly-Pro-pNAhydrochlorIde 10mg
SDP
Catalog No. 502981226
Encompass_Preferred
A chromogenic substrate that can be cleaved by DPP IV; enzyme activity can be quantified by colorimetric detection of free p-nitroanilide at 405 nm
Catalog No. 50-298-1226 Supplier Cayman Chemical Supplier No. 2124410 MG
A chromogenic substrate that can be cleaved by DPP IV; enzyme activity can be quantified by colorimetric detection of free p-nitroanilide at 405 nm