Learn More
Agilent Technologies GLY-X VACUUM MANIFOLD SPACER
Catalog No. NC1395653
Encompass
Gly-X Vacuum Manifold Spacer (2-pack) The Gly-X Vacuum Manifold Spacer ensures precise fitment of Gly-X components in the recommended Millipore MSVMHTS00-HTS vacuum manifold. Includes 2 reusable spacers.
Catalog No. NC1395653 Supplier Agilent Technologies Supplier No. GX100
Gly-X Vacuum Manifold Spacer (2-pack) The Gly-X Vacuum Manifold Spacer ensures precise fitment of Gly-X components in the recommended Millipore MSVMHTS00-HTS vacuum manifold. Includes 2 reusable spacers.