Learn More
Echelon Biosciences Research Labs [Gly22] Beta Amyloid (1-40), human - 0.5 mg, CAS#: 175010-18-1
SDP
Supplier:  Echelon Biosciences Research Labs 641140.5MG
Encompass_Preferred
[Gly22] Beta Amyloid (1-40), human - 0.5mg , Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gly-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH Category/Application: Alzheimer's This item is non returnable
Catalog No. 50-190-8687