Learn More
Medchemexpress LLC HY-131056 10mg , Gly3-VC-PAB-MMAE CAS:2684216-48-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13105610MG
Encompass_Preferred
HY-131056 10mg Gly3-VC-PAB-MMAE CAS: 2684216-48-4 Gly3-VC-PAB-MMAE consists a cleavable ADC linker (Gly3-VC-PAB) and a potent tubulin inhibitor (MMAE). Gly3-VC-PAB-MMAE can be used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0710