Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glyasperin C | 142474-53-1 | 98.9% | 356.41 | 1 MG
SDP

Catalog No. 5000426446
Encompass_Preferred

Glyasperin C is a potent and reversible noncompetitive inhibitor of neuraminidase with an IC50 of 20% at 200 μM. It is a polyphenol isolated from the roots of *Glycyrrhiza uralensis*.

  • Chemical formula: C21H24O5
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants, Leguminosae, *Glycyrrhiza uralensis* Fisch.
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed, away from moisture and light
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light)
  • Solubility in vitro: DMSO : 100 mg/mL (280.58 mM); requires ultrasonic

Catalog No. 50-004-26446 Supplier Medchemexpress LLC Supplier No. HYN99191MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glyasperin C is a potent and reversible noncompetitive inhibitor of neuraminidase with an IC50 of 20% at 200 μM. It is a polyphenol isolated from the roots of *Glycyrrhiza uralensis*.

  • Chemical formula: C21H24O5
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Plants, Leguminosae, *Glycyrrhiza uralensis* Fisch.
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed, away from moisture and light
  • Storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light)
  • Solubility in vitro: DMSO : 100 mg/mL (280.58 mM); requires ultrasonic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.