Learn More
Aviva Systems Biology Corp GLYAT Antibody - N-terminal region (ARP48682_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLYAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLYAT. Alternate names: ACGNAT, CAT, GATApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59884