Learn More
ABclonal Technology GLYAT Rabbit pAb
SDP
Catalog No. 5000109022
Encompass_Preferred
The glycine-N-acyltransferase protein conjugates glycine with acyl-CoA substrates in the mitochondria. The protein is thought to be important in the detoxification of endogenous and xenobiotic acyl-CoAs. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-09022 Supplier ABclonal Technology Supplier No. A14100500UL
The glycine-N-acyltransferase protein conjugates glycine with acyl-CoA substrates in the mitochondria. The protein is thought to be important in the detoxification of endogenous and xenobiotic acyl-CoAs. Two transcript variants encoding different isoforms have been found for this gene.